Product Details
- SNP ID
-
rs2822529
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:14277351 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTACATAGAAATTACTATGTGCA[A/G]TATATAATCAACTCAATGAGATCCA
- Phenotype
-
MIM: 608835
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs376138016] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCC13
- Gene Name
- ATP binding cassette subfamily C member 13 (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details