Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001270467.1 | 276 | Intron | NP_001257396.1 | ||
NM_001270468.1 | 276 | Intron | NP_001257397.1 | ||
NM_001270469.1 | 276 | Intron | NP_001257398.1 | ||
NM_001270470.1 | 276 | Intron | NP_001257399.1 | ||
NM_001270471.1 | 276 | Intron | NP_001257400.1 | ||
NM_003877.4 | 276 | UTR 5 | NP_003868.1 | ||
XM_011538929.1 | 276 | Intron | XP_011537231.1 | ||
XM_011538935.1 | 276 | Intron | XP_011537237.1 | ||
XM_011538936.1 | 276 | Intron | XP_011537238.1 | ||
XM_017020147.1 | 276 | UTR 5 | XP_016875636.1 | ||
XM_017020148.1 | 276 | Intron | XP_016875637.1 | ||
XM_017020149.1 | 276 | Intron | XP_016875638.1 | ||
XM_017020150.1 | 276 | Intron | XP_016875639.1 | ||
XM_017020151.1 | 276 | Intron | XP_016875640.1 | ||
XM_017020152.1 | 276 | Intron | XP_016875641.1 | ||
XM_017020153.1 | 276 | Intron | XP_016875642.1 | ||
XM_017020154.1 | 276 | Intron | XP_016875643.1 | ||
XM_017020155.1 | 276 | Intron | XP_016875644.1 | ||
XM_017020156.1 | 276 | Intron | XP_016875645.1 |