Product Details

SNP ID
rs2241835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24947948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATATAACACAAATTGAAAAAGT[C/T]ATTATTGCACAATGCAAAAAGTATA
Phenotype
MIM: 613527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC27 PubMed Links

Gene Details

Gene
DNAJC27
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198559.1 Intron NP_001185488.1
NM_016544.2 Intron NP_057628.1
XM_011532902.1 Intron XP_011531204.1
XM_017004269.1 Intron XP_016859758.1
XM_017004270.1 Intron XP_016859759.1

View Full Product Details