Product Details

SNP ID
rs2241837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24947649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTTCAGGAGGGCTGTCCGAGC[A/G]TTCACAACTGCTTTGAAGGCATCTT
Phenotype
MIM: 613527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC27 PubMed Links

Gene Details

Gene
DNAJC27
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198559.1 870 UTR 3 NP_001185488.1
NM_016544.2 870 Silent Mutation AAC,AAT N263N NP_057628.1
XM_011532902.1 870 Intron XP_011531204.1
XM_017004269.1 870 Silent Mutation AAC,AAT N192N XP_016859758.1
XM_017004270.1 870 Silent Mutation AAC,AAT N192N XP_016859759.1

View Full Product Details