Product Details

SNP ID
rs2285827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2967271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCAGCTCGGCGCGCCCTGCCC[G/T]CTGTTCCCACCCCGCTCTCCCCACC
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253725.1 Intron NP_001240654.1
NM_001253726.1 Intron NP_001240655.1
NM_024507.3 Intron NP_078783.1
NM_172229.2 Intron NP_757384.1
Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
There are no transcripts associated with this gene.

Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
There are no transcripts associated with this gene.

View Full Product Details