Product Details

SNP ID
rs2286242
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:43975683 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TACACAGCTGTACTGTCACCTGGGG[A/G]CAGCCTGAGGACGCCCCTTCAGGGA
Phenotype
MIM: 603717 MIM: 604013 MIM: 603456 MIM: 610411
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0B PubMed Links

Gene Details

Gene
ATP6V0B
Gene Name
ATPase H+ transporting V0 subunit b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039457.2 Intron NP_001034546.1
NM_001294333.1 Intron NP_001281262.1
NM_004047.4 Intron NP_004038.1
Gene
B4GALT2
Gene Name
beta-1,4-galactosyltransferase 2
There are no transcripts associated with this gene.

Gene
DPH2
Gene Name
DPH2 homolog
There are no transcripts associated with this gene.

Gene
IPO13
Gene Name
importin 13
There are no transcripts associated with this gene.

View Full Product Details