Product Details
- SNP ID
-
rs2287359
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89179482 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCACCCTGTACCCCTGGCGCCGG[G/T]CGCCTGCCCTGAGCCGCGTGCGGAG
- Phenotype
-
MIM: 114019
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDH15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76549400] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDH15
- Gene Name
- cadherin 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004933.2 |
186 |
Missense Mutation |
GCG,TCG |
A37S |
NP_004924.1 |
View Full Product Details