Product Details

SNP ID
rs2307469
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:37109260 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAGCAAGAATTAGCCCCAAAGCGTA[C/G]AGGTCCACTTCCTTTCCATAGTCTT
Phenotype
MIM: 176871
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF2AK2 PubMed Links

Gene Details

Gene
EIF2AK2
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135651.2 1670 Silent Mutation CTC,CTG L471L NP_001129123.1
NM_001135652.2 1670 Silent Mutation CTC,CTG L430L NP_001129124.1
NM_002759.3 1670 Silent Mutation CTC,CTG L471L NP_002750.1
XM_011532987.2 1670 Silent Mutation CTC,CTG L471L XP_011531289.1
XM_017004503.1 1670 Silent Mutation CTC,CTG L428L XP_016859992.1

View Full Product Details