Product Details

SNP ID
rs2072313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15936108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTAGAGGGGGTGGTCTCTTTGCC[C/T]TTGATGTGCTCCCTGGGGGACATCT
Phenotype
MIM: 607737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFBP1 PubMed Links

Gene Details

Gene
FGFBP1
Gene Name
fibroblast growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005130.4 820 Silent Mutation AAA,AAG K175K NP_005121.1

View Full Product Details