Product Details

SNP ID
rs11669315
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:15728649 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCTGTGGCCCTGGGCGTGGGCTT[A/G]GTATGTATCATGGCACTGCTGGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10H2 PubMed Links

Gene Details

Gene
OR10H2
Gene Name
olfactory receptor family 10 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013939.2 626 Silent Mutation TTA,TTG L202L NP_039227.1

View Full Product Details