Product Details

SNP ID
rs3748051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35797166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCAAGCTTAGGAAGTCTTCATC[C/T]GGGGACTCAGGAGGCGCCTCCCCTG
Phenotype
MIM: 120105 MIM: 609427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLPS PubMed Links

Gene Details

Gene
CLPS
Gene Name
colipase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252597.1 Intron NP_001239526.1
NM_001252598.1 Intron NP_001239527.1
NM_001832.3 Intron NP_001823.1
Gene
CLPSL1
Gene Name
colipase like 1
There are no transcripts associated with this gene.

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
There are no transcripts associated with this gene.

View Full Product Details