Product Details

SNP ID
rs116387840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106925785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAGAGGCCCATTAACACTTA[G/T]TGAGATCTTTTACCATTATTGCAGC
Phenotype
MIM: 605415
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DKK2 PubMed Links

Gene Details

Gene
DKK2
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014421.2 Intron NP_055236.1

View Full Product Details