Product Details
- SNP ID
-
rs17118135
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:55939426 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTGCCTTCCTAGACTCTGAAGGA[C/T]GACGGACAGCACATGTGGAGGCCCA
- Phenotype
-
MIM: 125855
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DGKA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113515918] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DGKA
- Gene Name
- diacylglycerol kinase alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001345.4 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
NP_001336.2 |
NM_201444.2 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
NP_958852.1 |
NM_201445.1 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
NP_958853.1 |
NM_201554.1 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
NP_963848.1 |
XM_005268688.2 |
701 |
Silent Mutation |
GAC,GAT |
D240D |
XP_005268745.1 |
XM_005268689.2 |
701 |
Silent Mutation |
GAC,GAT |
D90D |
XP_005268746.1 |
XM_005268690.2 |
701 |
Silent Mutation |
GAC,GAT |
D90D |
XP_005268747.1 |
XM_011537991.2 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
XP_011536293.1 |
XM_011537993.2 |
701 |
Silent Mutation |
GAC,GAT |
D202D |
XP_011536295.1 |
XM_011537995.2 |
701 |
Silent Mutation |
GAC,GAT |
D90D |
XP_011536297.1 |
XM_017018900.1 |
701 |
Silent Mutation |
GAC,GAT |
D358D |
XP_016874389.1 |
XM_017018901.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874390.1 |
XM_017018902.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874391.1 |
XM_017018903.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874392.1 |
XM_017018904.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874393.1 |
XM_017018905.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874394.1 |
XM_017018906.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874395.1 |
XM_017018907.1 |
701 |
Silent Mutation |
GAC,GAT |
D320D |
XP_016874396.1 |
XM_017018908.1 |
701 |
Silent Mutation |
GAC,GAT |
D286D |
XP_016874397.1 |
XM_017018909.1 |
701 |
Silent Mutation |
GAC,GAT |
D61D |
XP_016874398.1 |
View Full Product Details