Product Details

SNP ID
rs3133187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75399545 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCTTTCAGCGGAGCGCGGCGGC[A/G]AGATGGCAGTGCAAATATCCAAGAA
Phenotype
MIM: 600454 MIM: 600455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS3 PubMed Links

Gene Details

Gene
RPS3
Gene Name
ribosomal protein S3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005.4 55 UTR 5 NP_000996.2
NM_001256802.1 55 UTR 5 NP_001243731.1
NM_001260506.1 55 UTR 5 NP_001247435.1
NM_001260507.1 55 UTR 5 NP_001247436.1
Gene
SNORD15A
Gene Name
small nucleolar RNA, C/D box 15A
There are no transcripts associated with this gene.

Gene
SNORD15B
Gene Name
small nucleolar RNA, C/D box 15B
There are no transcripts associated with this gene.

View Full Product Details