Product Details

SNP ID
rs3739473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:78302180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCAATTCACTATTAGTACCTAA[C/T]GCTTTTTATTTGTTCCCCATCCCGT
Phenotype
MIM: 610936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSAT1 PubMed Links

Gene Details

Gene
PSAT1
Gene Name
phosphoserine aminotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021154.4 Intron NP_066977.1
NM_058179.3 Intron NP_478059.1

View Full Product Details