Product Details

SNP ID
rs112785437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35737039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCAACCCCCATTTACTGGCACA[A/G]TTGGGTGGAGAGAAGGGAAGGGGTA
Phenotype
MIM: 606443 MIM: 609471 MIM: 186745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB3 PubMed Links

Gene Details

Gene
CREB3
Gene Name
cAMP responsive element binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006368.4 3417 Intron NP_006359.3
Gene
GBA2
Gene Name
glucosylceramidase beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020944.2 3417 UTR 3 NP_065995.1
XM_005251526.4 3417 UTR 3 XP_005251583.1
XM_006716809.3 3417 UTR 3 XP_006716872.1
XM_011517973.2 3417 UTR 3 XP_011516275.1
XM_017014937.1 3417 UTR 3 XP_016870426.1
XM_017014938.1 3417 UTR 3 XP_016870427.1
XM_017014939.1 3417 UTR 3 XP_016870428.1
XM_017014940.1 3417 UTR 3 XP_016870429.1
XM_017014941.1 3417 Intron XP_016870430.1
XM_017014942.1 3417 UTR 3 XP_016870431.1
XM_017014943.1 3417 UTR 3 XP_016870432.1
XM_017014944.1 3417 UTR 3 XP_016870433.1
XM_017014945.1 3417 UTR 3 XP_016870434.1
XM_017014946.1 3417 UTR 3 XP_016870435.1
Gene
MIR6853
Gene Name
microRNA 6853
There are no transcripts associated with this gene.

Gene
TLN1
Gene Name
talin 1
There are no transcripts associated with this gene.

View Full Product Details