Product Details

SNP ID
rs1007050688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134383579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCTCCCAGCCCGGCTTCCCGGG[A/T]TCCCCCTGCGCGCGCCTCCGCACCC
Phenotype
MIM: 151290
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GAT1 PubMed Links

Gene Details

Gene
B3GAT1
Gene Name
beta-1,3-glucuronyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018644.3 Intron NP_061114.2
NM_054025.2 Intron NP_473366.1
Gene
GLB1L2
Gene Name
galactosidase beta 1 like 2
There are no transcripts associated with this gene.

View Full Product Details