Product Details

SNP ID
hCV25619489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:114570811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAATAGTACCAATTAAAAGTGAA[C/T]TTTTTCAAGTTTTTCTGCCAAAAAT
Phenotype
MIM: 605783
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005872.2 Intron NP_005863.1

View Full Product Details