Product Details

SNP ID
rs74124228
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:172532716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACCATGGATTCTAGGAGGGAC[C/T]GGGAAAGAGAAAAACGAATTCTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 368 Intron NP_001269679.1
NM_001282751.1 368 Intron NP_001269680.1
NM_014283.4 368 Intron NP_055098.1
NM_016227.3 368 Missense Mutation CGG,TGG R82W NP_057311.3
XM_006711374.2 368 Intron XP_006711437.1
XM_006711375.2 368 Intron XP_006711438.1
XM_006711376.1 368 Intron XP_006711439.1

View Full Product Details