Product Details

SNP ID
rs61733387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107872813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGTCACTATTCTTACCTTCA[A/G]TGGTTTAATCTGATCCAAACCCATA
Phenotype
MIM: 608648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 2152 Silent Mutation CTG,TTG L712L NP_009145.1
XM_011535399.2 2152 Silent Mutation CTG,TTG L656L XP_011533701.1
XM_017010218.1 2152 Silent Mutation CTG,TTG L346L XP_016865707.1

View Full Product Details