Product Details

SNP ID
rs62636580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151822799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGATCTTCTTGGCCCTCTGGAT[A/G]AAGAGTTTTCGCATCTCCTCTGGGG
Phenotype
MIM: 138491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLRA1 PubMed Links

Gene Details

Gene
GLRA1
Gene Name
glycine receptor alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000171.3 1511 Silent Mutation TTC,TTT F408F NP_000162.2
NM_001146040.1 1511 Silent Mutation TTC,TTT F416F NP_001139512.1
NM_001292000.1 1511 Silent Mutation TTC,TTT F325F NP_001278929.1
XM_017009349.1 1511 Silent Mutation TTC,TTT F424F XP_016864838.1

View Full Product Details