Product Details
- SNP ID
-
rs35901190
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37056357 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCGAGTTCTACAACATCGGCCC[A/G]CTGATCCGCATCATCAAAGACCGGA
- Phenotype
-
MIM: 616386
MIM: 609862
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD17
PubMed Links
Gene Details
- Gene
- KCTD17
- Gene Name
- potassium channel tetramerization domain containing 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282684.1 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
NP_001269613.1 |
NM_001282685.1 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
NP_001269614.1 |
NM_001282686.1 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
NP_001269615.1 |
NM_024681.3 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
NP_078957.2 |
XM_005261741.3 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_005261798.2 |
XM_005261742.2 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_005261799.2 |
XM_005261743.2 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_005261800.2 |
XM_005261744.2 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_005261801.1 |
XM_011530374.2 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_011528676.1 |
XM_011530377.2 |
372 |
Silent Mutation |
CCA,CCG |
P119P |
XP_011528679.1 |
- Gene
- TMPRSS6
- Gene Name
- transmembrane protease, serine 6
There are no transcripts associated with this gene.
View Full Product Details