Product Details

SNP ID
rs11551619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42799103 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCCAGCAACCGATCTCACTCCC[C/T]GCTTGAACTGCGCCATGTCGGGGGC
Phenotype
MIM: 612439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP3 PubMed Links

Gene Details

Gene
ARFGAP3
Gene Name
ADP ribosylation factor GTPase activating protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142293.1 1689 Missense Mutation CAG,CGG Q446R NP_001135765.1
NM_014570.4 1689 Missense Mutation CAG,CGG Q490R NP_055385.3
XM_005261525.3 1689 Missense Mutation CAG,CGG Q446R XP_005261582.1

View Full Product Details