Product Details

SNP ID
rs56231264
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:15511563 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTTGAAAGAGAAAGGTCAAAAAAAG[C/T]CATAAAAGAGAGTCGTTTTTTGAGG
Phenotype
MIM: 300335 MIM: 300101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACE2 PubMed Links

Gene Details

Gene
ACE2
Gene Name
angiotensin I converting enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021804.2 Intron NP_068576.1
XM_011545549.1 Intron XP_011543851.1
XM_011545551.2 Intron XP_011543853.1
XM_011545552.1 Intron XP_011543854.1
Gene
BMX
Gene Name
BMX non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320866.1 Intron NP_001307795.1
NM_001721.6 Intron NP_001712.1
NM_203281.2 Intron NP_975010.1
XM_017029752.1 Intron XP_016885241.1

View Full Product Details