Product Details

SNP ID
rs61732784
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:1501394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGCTGGAGACTTGGTCCAGCC[A/G]GGCATCAGCTTTCCGGGGCCGGCAG
Phenotype
MIM: 605438
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DLGAP2 PubMed Links

Gene Details

Gene
DLGAP2
Gene Name
DLG associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277161.1 29 UTR 5 NP_001264090.1
NM_004745.4 29 UTR 5 NP_004736.2

View Full Product Details