Product Details
- SNP ID
-
rs17088860
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:74149176 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACAAGAGCTGCCAATGCCCTTTTA[C/T]ACCAAGTTCACTGCTAAAGGTGAAA
- Phenotype
-
MIM: 609093
MIM: 615180
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112469115] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO15
- Gene Name
- F-box protein 15
There are no transcripts associated with this gene.
- Gene
- TIMM21
- Gene Name
- translocase of inner mitochondrial membrane 21
View Full Product Details