Product Details

SNP ID
rs73991016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219077145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCACAAATATCCCTCAATATTT[C/T]TGAGGCAGTGCCAATGAAGCAGCAA
Phenotype
MIM: 611290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHEJ1 PubMed Links

Gene Details

Gene
NHEJ1
Gene Name
non-homologous end joining factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024782.2 Intron NP_079058.1

View Full Product Details