Product Details

SNP ID
rs35332743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7580306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATTCAGATTCGAGTCATGTA[C/T]ACAACCCAGGGTGGAGGAGAGGTAA
Phenotype
MIM: 153634 MIM: 602641 MIM: 604041 MIM: 612844 MIM: 601297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD68 PubMed Links

Gene Details

Gene
CD68
Gene Name
CD68 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040059.1 737 Silent Mutation TAC,TAT Y155Y NP_001035148.1
NM_001251.2 737 Silent Mutation TAC,TAT Y182Y NP_001242.2
Gene
EIF4A1
Gene Name
eukaryotic translation initiation factor 4A1
There are no transcripts associated with this gene.

Gene
LOC100996842
Gene Name
uncharacterized LOC100996842
There are no transcripts associated with this gene.

Gene
MPDU1
Gene Name
mannose-P-dolichol utilization defect 1
There are no transcripts associated with this gene.

Gene
SENP3
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 3
There are no transcripts associated with this gene.

Gene
SENP3-EIF4A1
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SNORA48
Gene Name
small nucleolar RNA, H/ACA box 48
There are no transcripts associated with this gene.

Gene
SNORA67
Gene Name
small nucleolar RNA, H/ACA box 67
There are no transcripts associated with this gene.

Gene
SNORD10
Gene Name
small nucleolar RNA, C/D box 10
There are no transcripts associated with this gene.

Gene
SOX15
Gene Name
SRY-box 15
There are no transcripts associated with this gene.

View Full Product Details