Product Details

SNP ID
rs74103032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68748155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATAGACTGTTGGGAAGAAAGC[A/C]TAATTGCATAAGTTGTCTGACTCAA
Phenotype
MIM: 607617
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUP107 PubMed Links

Gene Details

Gene
NUP107
Gene Name
nucleoporin 107
There are no transcripts associated with this gene.

Gene
SLC35E3
Gene Name
solute carrier family 35 member E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018656.2 Intron NP_061126.2
XM_005269006.3 Intron XP_005269063.1
XM_017019584.1 Intron XP_016875073.1

View Full Product Details