Product Details

SNP ID
rs61743809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68746441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGCCGCCGGGCTCCTGTTCAAC[C/T]TGCTGGTGTCCATCTGCATTGTGTT
Phenotype
MIM: 607617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP107 PubMed Links

Gene Details

Gene
NUP107
Gene Name
nucleoporin 107
There are no transcripts associated with this gene.

Gene
SLC35E3
Gene Name
solute carrier family 35 member E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018656.2 294 Silent Mutation CTG,TTG L22L NP_061126.2
XM_005269006.3 294 Missense Mutation CTG,TTG L22L XP_005269063.1
XM_017019584.1 294 Missense Mutation CTG,TTG L22L XP_016875073.1

View Full Product Details