Product Details

SNP ID
rs34154432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160677604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTGAGAACACCGATGGGCTGGT[A/G]TATGTTGTGGACAGTACAGACAAAC
Phenotype
MIM: 614439
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL14 PubMed Links

Gene Details

Gene
ARL14
Gene Name
ADP ribosylation factor like GTPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025047.2 445 Silent Mutation GTA,GTG V86V NP_079323.1

View Full Product Details