Product Details

SNP ID
rs61746514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149983293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGGCGAGGCCTGCCTAGCGGGC[C/T]GGGCCCCCGGGCCATGTCTCCGGCC
Phenotype
MIM: 611365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM183B PubMed Links

Gene Details

Gene
TMEM183B
Gene Name
transmembrane protein 183B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079809.1 Intron NP_001073277.1

View Full Product Details