Product Details

SNP ID
rs35182771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:93829637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGCAAAAGCGACCACCACCA[A/C]CTTCGCACCGCGGCCTCCTATGAAG
Phenotype
MIM: 601461
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATOH1 PubMed Links

Gene Details

Gene
ATOH1
Gene Name
atonal bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005172.1 711 Silent Mutation CAA,CAC Q237H NP_005163.1

View Full Product Details