Product Details

SNP ID
rs34362649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9887880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGCATCACTGAAGGAGGCCGG[A/G]GAGAGGCTGGTGGCTGTGGACTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 1850 Silent Mutation GGA,GGG G467G NP_001091999.1
NM_032243.5 1850 Silent Mutation GGA,GGG G400G NP_115619.4
XM_017026041.1 1850 Silent Mutation GGA,GGG G400G XP_016881530.1

View Full Product Details