Product Details

SNP ID
rs61730681
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:64190230 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTGTAGGTGTGAGGGACGAGCTAG[C/T]TGAGGCCCAGATTCCCCAGGTGCCC
Phenotype
MIM: 300647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 3330 Silent Mutation CAA,CAG Q1019Q NP_689637.3

View Full Product Details