Product Details

SNP ID
rs73633794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57351296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTTTCCTATGCCCGGTCCGTG[C/T]ATTCTGGTTGTGAAGGCTGAGTTCT
Phenotype
MIM: 613840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF304 PubMed Links

Gene Details

Gene
ZNF304
Gene Name
zinc finger protein 304
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290318.1 23 UTR 5 NP_001277247.1
NM_001290319.1 23 UTR 5 NP_001277248.1
NM_020657.3 23 UTR 5 NP_065708.2
XM_011527145.2 23 UTR 5 XP_011525447.1
XM_017027024.1 23 UTR 5 XP_016882513.1

View Full Product Details