Product Details

SNP ID
rs2074071
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57492212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGTGACTTCAGCCATACCGAGAG[A/G]TAGTTGGTGGGTGGAGCTCAGGGAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF419 PubMed Links

Gene Details

Gene
ZNF419
Gene Name
zinc finger protein 419
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098491.1 Intron NP_001091961.1
NM_001098492.1 Intron NP_001091962.1
NM_001098493.1 Intron NP_001091963.1
NM_001098494.1 Intron NP_001091964.1
NM_001098495.1 Intron NP_001091965.1
NM_001098496.1 Intron NP_001091966.1
NM_001291743.1 Intron NP_001278672.1
NM_001291744.1 Intron NP_001278673.1
NM_001291745.1 Intron NP_001278674.1
NM_024691.3 Intron NP_078967.3
Gene
ZNF773
Gene Name
zinc finger protein 773
There are no transcripts associated with this gene.

View Full Product Details