Product Details

SNP ID
rs115574954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22224186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCAGGAAGGGCTGGGAGGGAG[A/G]AGAGGCCCGGCGGGTCCAGCCGGGA
Phenotype
MIM: 608511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHYHIP PubMed Links

Gene Details

Gene
PHYHIP
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099335.1 Intron NP_001092805.1
NM_014759.3 Intron NP_055574.3
XM_006716416.1 Intron XP_006716479.1
XM_017014102.1 Intron XP_016869591.1

View Full Product Details