Product Details
- SNP ID
-
rs867950775
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41911109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTCACCCGCAGAGGCCGCGGCTC[A/T]TTTCCACCGGGGAAGCCTCTATGCC
- Phenotype
-
MIM: 600481
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76140449] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHISA8
- Gene Name
- shisa family member 8
- Gene
- SREBF2
- Gene Name
- sterol regulatory element binding transcription factor 2
There are no transcripts associated with this gene.
View Full Product Details