Product Details

SNP ID
rs3766163
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:15660052 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAAGGCTTTGAAGGCTTCAGCTGAA[C/T]TCCAGCTAAACTCTGAAAAGAAAGA
Phenotype
MIM: 601966
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDI2 PubMed Links

Gene Details

Gene
DDI2
Gene Name
DNA damage inducible 1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032341.4 340 UTR 3 NP_115717.3
Gene
RSC1A1
Gene Name
regulatory solute carrier protein, family 1, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006511.2 340 Missense Mutation CTC,TTC L62F NP_006502.1

View Full Product Details