Product Details

SNP ID
rs7103540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4822024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATAGGATCAACAGCATCCTTGG[A/T]CTGTTTGCGCTTTTGTCCACTACAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR51F2 PubMed Links

Gene Details

Gene
OR51F2
Gene Name
olfactory receptor family 51 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004753.1 639 Silent Mutation GGA,GGT G213G NP_001004753.1

View Full Product Details