Product Details

SNP ID
hCV25771762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36638393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATTTCACTGTTTCAGTGAAAT[A/G]TTTCACTGGTTTCAGTGAAACATGT
Phenotype
MIM: 609516 MIM: 608640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF382 PubMed Links

Gene Details

Gene
ZNF382
Gene Name
zinc finger protein 382
There are no transcripts associated with this gene.

Gene
ZNF461
Gene Name
zinc finger protein 461
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297623.2 2159 UTR 3 NP_001284552.1
NM_001322821.1 2159 UTR 3 NP_001309750.1
NM_001322823.1 2159 UTR 3 NP_001309752.1
NM_001322825.1 2159 UTR 3 NP_001309754.1
NM_001322826.1 2159 UTR 3 NP_001309755.1
NM_001322827.1 2159 UTR 3 NP_001309756.1
NM_001322828.1 2159 UTR 3 NP_001309757.1
NM_153257.4 2159 UTR 3 NP_694989.2

View Full Product Details