Product Details
- SNP ID
-
rs45458093
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29792759 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTACCCCTCCACATCCCCCACG[A/G]ACCGCCCGGGTCTCCCCGAGTCTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HCG4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192929095] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCG4
- Gene Name
- HLA complex group 4 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- LOC554223
- Gene Name
- histocompatibility antigen-related
There are no transcripts associated with this gene.
View Full Product Details