Product Details
- SNP ID
-
rs16823066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:169482299 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTCCTTATTGATTGTTTAGATTC[C/T]ATTGAAGACACCAAAGGAAATAATG
- Phenotype
-
MIM: 603650
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BBS5
PubMed Links
Gene Details
- Gene
- BBS5
- Gene Name
- Bardet-Biedl syndrome 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152384.2 |
166 |
Silent Mutation |
TCC,TCT |
S36S |
NP_689597.1 |
View Full Product Details