Product Details

SNP ID
rs56222810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25012584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCAGGAATCAGGGAGGAGCTC[A/G]TCTCCAGGACACTCACCTTCTCCTT
Phenotype
MIM: 614355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSS1 PubMed Links

Gene Details

Gene
ACSS1
Gene Name
acyl-CoA synthetase short-chain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252675.1 Intron NP_001239604.1
NM_001252676.1 Intron NP_001239605.1
NM_001252677.1 Intron NP_001239606.1
NM_032501.3 Intron NP_115890.2
XM_006723659.1 Intron XP_006723722.1
XM_011529388.1 Intron XP_011527690.1

View Full Product Details