Product Details

SNP ID
rs10082935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107319103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGGCGGGAGCTGCTGGCCGCCT[C/T]TAGCTGACTCCATGCACAGCCGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD11 PubMed Links

Gene Details

Gene
BTBD11
Gene Name
BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017523.1 1936 Intron NP_001017523.1
NM_001018072.1 1936 Silent Mutation CTA,TTA L55L NP_001018082.1
XM_005268645.3 1936 Intron XP_005268702.1
XM_011537909.2 1936 Intron XP_011536211.1
XM_011537910.2 1936 Intron XP_011536212.1
XM_017018807.1 1936 Silent Mutation CTA,TTA L55L XP_016874296.1

View Full Product Details