Product Details

SNP ID
rs61738797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1349550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCTGCTGCTGGCTCCTCCGC[A/G]CTCGGCGCCAGGCCCTGGGTCCTCA
Phenotype
MIM: 604009 MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199096.1 894 Intron NP_001186025.1
NM_001199097.1 894 Intron NP_001186026.1
NM_001199098.1 894 Intron NP_001186027.1
NM_001199099.1 894 Intron NP_001186028.1
NM_001286464.1 894 Intron NP_001273393.1
NM_003933.4 894 Intron NP_003924.2
XM_011522728.1 894 Intron XP_011521030.1
XM_011522729.1 894 Intron XP_011521031.1
XM_011522730.2 894 Intron XP_011521032.1
Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
There are no transcripts associated with this gene.

Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001410.2 894 Missense Mutation CGC,TGC R276C NP_001001410.1

View Full Product Details