Product Details

SNP ID
rs35891903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:648943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATTTTCTCTTTATCTGCCGG[G/T]GGGGGGCCTGTGTTTTCAAACTGAT
Phenotype
MIM: 605831 MIM: 615177
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGF22 PubMed Links

Gene Details

Gene
FGF22
Gene Name
fibroblast growth factor 22
There are no transcripts associated with this gene.

Gene
RNF126
Gene Name
ring finger protein 126
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194460.2 720 Intron NP_919442.1
XM_005259594.1 720 Silent Mutation CCA,CCC P176P XP_005259651.1

View Full Product Details