Product Details
- SNP ID
-
rs16996376
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19795322 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGCATCAATTTTCTTATATGTAG[G/T]ACGGGTTGAAGACTGGTTAAAAGTT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF506
PubMed Links
Gene Details
- Gene
- ZNF506
- Gene Name
- zinc finger protein 506
View Full Product Details