Product Details

SNP ID
rs76343572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30784535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTACTGGTAACACAGCTCATTTC[C/G]TAATTCAGATTTTAAAAAATCTGGA
Phenotype
MIM: 614853
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRLF3 PubMed Links

Gene Details

Gene
CRLF3
Gene Name
cytokine receptor like factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015986.3 Intron NP_057070.3
XM_011524879.1 Intron XP_011523181.1

View Full Product Details